Product Details

SNP ID
rs201425251
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:39897596 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACGCAGGTCATTCCGCCTCTTGC[A/G]CTCCAGGAAGTTGTGATTCTTCCTC
Phenotype
MIM: 164850
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYCL PubMed Links

Gene Details

Gene
MYCL
Gene Name
v-myc avian myelocytomatosis viral oncogene lung carcinoma derived homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033081.2 1452 Missense Mutation CGC,TGC R291C NP_001028253.1
NM_001033082.2 1452 Missense Mutation CGC,TGC R321C NP_001028254.2
NM_005376.4 1452 Intron NP_005367.2

View Full Product Details