Product Details

SNP ID
rs201583978
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:11273761 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGCCCTTGCCTACAGATCCCACG[A/G]TGTCCTGGATCCCAGGCTCTTGGTG
Phenotype
MIM: 611632
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UBIAD1 PubMed Links

Gene Details

Gene
UBIAD1
Gene Name
UbiA prenyltransferase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013319.2 564 Missense Mutation GAT,GGT D77G NP_037451.1
XM_006710590.3 564 Missense Mutation GAT,GGT D77G XP_006710653.1
XM_011541304.2 564 Missense Mutation GAT,GGT D77G XP_011539606.1

View Full Product Details