Product Details

SNP ID
rs199716905
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:1203782 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGAGCAGAACGCAGAGCCCCTG[C/T]GGCCTGGGGAGCTCCTGGGGAGGGG
Phenotype
MIM: 603905 MIM: 600315
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TNFRSF18 PubMed Links

Gene Details

Gene
TNFRSF18
Gene Name
TNF receptor superfamily member 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004195.2 905 UTR 3 NP_004186.1
NM_148901.1 905 Missense Mutation ACA,GCA T193A NP_683699.1
NM_148902.1 905 UTR 3 NP_683700.1
XM_017002722.1 905 Missense Mutation ACA,GCA T285A XP_016858211.1
Gene
TNFRSF4
Gene Name
TNF receptor superfamily member 4
There are no transcripts associated with this gene.

Gene
TTLL10
Gene Name
tubulin tyrosine ligase like 10
There are no transcripts associated with this gene.

View Full Product Details