Product Details
- SNP ID
-
rs199785140
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:232805375 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGCAGGCAGCGATGCCTTTTGCTC[C/T]GGTTTCTCGTGAGACCCCGGGGCTT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MAP10
PubMed Links
Gene Details
- Gene
- MAP10
- Gene Name
- microtubule associated protein 10
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_019090.2 |
484 |
Missense Mutation |
CGG,TGG |
R118W |
NP_061963.2 |
View Full Product Details