Product Details

SNP ID
rs199536952
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:15466137 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGGGGTGATGGCGTGATCTCCA[C/G]CTGCAACGTGAGTACCAAAATCAGG
Phenotype
MIM: 609443 MIM: 609444
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CELA2A PubMed Links

Gene Details

Gene
CELA2A
Gene Name
chymotrypsin like elastase family member 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033440.2 658 Missense Mutation ACC,AGC T211S NP_254275.1
Gene
CELA2B
Gene Name
chymotrypsin like elastase family member 2B
There are no transcripts associated with this gene.

View Full Product Details