Product Details

SNP ID
rs199722325
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:207054756 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCTCAGAACAGAACAACAACAG[C/T]TATGAAACCAAAACCCCAAATCTTC
Phenotype
MIM: 171835 MIM: 612023
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PFKFB2 PubMed Links

Gene Details

Gene
PFKFB2
Gene Name
6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018053.1 148 Silent Mutation AGC,AGT S13S NP_001018063.1
NM_006212.2 148 Silent Mutation AGC,AGT S13S NP_006203.2
XM_005273162.3 148 Intron XP_005273219.1
Gene
YOD1
Gene Name
YOD1 deubiquitinase
There are no transcripts associated with this gene.

View Full Product Details