Product Details

SNP ID
rs200081331
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:109712654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCGCTTGCTCCTGGAATACACA[G/T]ACTCAAGCTATGTGGAAAAGAAGTA
Phenotype
MIM: 138385
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GSTM5 PubMed Links

Gene Details

Gene
GSTM5
Gene Name
glutathione S-transferase mu 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000851.3 170 Missense Mutation GAC,TAC D25Y NP_000842.2
XM_005270784.4 170 Missense Mutation GAC,TAC D25Y XP_005270841.1
XM_005270785.4 170 Intron XP_005270842.1

View Full Product Details