Product Details
- SNP ID
-
rs200042425
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:183023811 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGCGGCGGCCGCCGCGGCGGGCT[C/G]TGCCCAGGCAGCCATGGACGAGTGC
- Phenotype
-
MIM: 150290
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LAMC1
PubMed Links
Gene Details
- Gene
- LAMC1
- Gene Name
- laminin subunit gamma 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002293.3 |
352 |
Missense Mutation |
TCT,TGT |
S32C |
NP_002284.3 |
View Full Product Details