Product Details
- SNP ID
-
rs199817559
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:230704236 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCATTGCTCAATTTTTGCAGGTTC[A/G]GCTCGGTGTGCAGAATGGCGGGCAG
- Phenotype
-
MIM: 106150
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AGT
PubMed Links
Gene Details
- Gene
- AGT
- Gene Name
- angiotensinogen
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000029.3 |
1734 |
Missense Mutation |
CCG,CTG |
P409L |
NP_000020.1 |
View Full Product Details