Product Details

SNP ID
rs199632500
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:247491427 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACCTACCACGGCTGTGTGGCCCA[A/C]CTCTACATCTACATGATGCTGGGCT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR2W5 PubMed Links

Gene Details

Gene
OR2W5
Gene Name
olfactory receptor family 2 subfamily W member 5 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004698.2 360 Silent Mutation CAA,CAC Q100H NP_001004698.1

View Full Product Details