Product Details

SNP ID
rs200007349
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:247712262 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCACGGGGGCCACAGAAGGACAGG[C/T]CACTGATGAGGGCTGTGGGCACTGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR6F1 PubMed Links

Gene Details

Gene
OR6F1
Gene Name
olfactory receptor family 6 subfamily F member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005286.1 494 Missense Mutation GAC,GGC D165G NP_001005286.1

View Full Product Details