Product Details

SNP ID
rs199715708
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:21823711 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGTCGACTTGGATGGAACCTCTG[C/T]GGCCCTCCCTGCAGTGGAACTGGGT
Phenotype
MIM: 142461
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HSPG2 PubMed Links

Gene Details

Gene
HSPG2
Gene Name
heparan sulfate proteoglycan 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291860.1 12988 Missense Mutation CAC,CGC H4304R NP_001278789.1
NM_005529.6 12988 Missense Mutation CAC,CGC H4303R NP_005520.4
XM_011541318.2 12988 Missense Mutation CAC,CGC H4486R XP_011539620.1
XM_017001120.1 12988 Missense Mutation CAC,CGC H4368R XP_016856609.1
XM_017001121.1 12988 Missense Mutation CAC,CGC H4351R XP_016856610.1
XM_017001122.1 12988 Missense Mutation CAC,CGC H4350R XP_016856611.1
Gene
LDLRAD2
Gene Name
low density lipoprotein receptor class A domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013693.2 12988 UTR 3 NP_001013715.2
XM_005245873.4 12988 Intron XP_005245930.1

View Full Product Details