Product Details

SNP ID
rs200671620
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:182056575 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGGCAAAGGCCTGGCCACACTGC[A/G]TGCAAGGAAAGGGCCTCTCGCCAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF648 PubMed Links

Gene Details

Gene
ZNF648
Gene Name
zinc finger protein 648
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009992.1 5577 Missense Mutation ACG,ATG T479M NP_001009992.1
XM_017000301.1 5577 Missense Mutation ACG,ATG T479M XP_016855790.1
XM_017000302.1 5577 Missense Mutation ACG,ATG T479M XP_016855791.1

View Full Product Details