Product Details

SNP ID
rs200634484
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:108698693 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACAAAACTGGAGTGGTTTTCTAC[C/T]TTGTTTCCAAGAATTCCAGTTCCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRPF38B PubMed Links

Gene Details

Gene
PRPF38B
Gene Name
pre-mRNA processing factor 38B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018061.2 435 Silent Mutation ACC,ACT T216T NP_060531.2
XM_011541687.2 435 Intron XP_011539989.1
XM_017001596.1 435 Silent Mutation ACC,ACT T71T XP_016857085.1
XM_017001597.1 435 Silent Mutation ACC,ACT T32T XP_016857086.1
XM_017001598.1 435 Silent Mutation ACC,ACT T19T XP_016857087.1

View Full Product Details