Product Details

SNP ID
rs200276576
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:1387374 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCCGGATTATCCGCCCGCTCCC[A/G]TGACCTGCTTCGAGAACGGGAAGAA
Phenotype
MIM: 613482
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCNL2 PubMed Links

Gene Details

Gene
CCNL2
Gene Name
cyclin L2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039577.3 1336 Intron NP_001034666.1
NM_001320153.1 1336 Missense Mutation CGG,TGG R252W NP_001307082.1
NM_001320155.1 1336 Missense Mutation CGG,TGG R252W NP_001307084.1
NM_030937.4 1336 Missense Mutation CGG,TGG R474W NP_112199.2
XM_011542216.2 1336 Missense Mutation CGG,TGG R507W XP_011540518.1
XM_011542219.2 1336 Missense Mutation CGG,TGG R343W XP_011540521.1
XM_011542221.2 1336 Missense Mutation CGG,TGG R252W XP_011540523.1
XM_017002420.1 1336 Missense Mutation CGG,TGG R352W XP_016857909.1
XM_017002421.1 1336 Missense Mutation CGG,TGG R343W XP_016857910.1
XM_017002422.1 1336 Missense Mutation CGG,TGG R252W XP_016857911.1
XM_017002423.1 1336 Intron XP_016857912.1

View Full Product Details