Product Details

SNP ID
rs200172924
Assay Type
Functionally Tested
NCBI dbSNP Submissions
6
Location
Chr.1:27394159 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTGGCGTGCTGGCAATGGCCTTT[C/A]CCGCCAGCATCGGCAGTCTACTGGC
Phenotype
MIM: 600241
Polymorphism
C/A, Transversion Substitution
Allele Nomenclature
Literature Links
GPR3 PubMed Links

Gene Details

Gene
GPR3
Gene Name
G protein-coupled receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005281.3 460 Missense Mutation ACC,CCC T121P NP_005272.1

View Full Product Details