Product Details

SNP ID
rs200717164
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:114674758 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGAGCTTCCAACTCACCTTTTT[A/T]AAATTTAGGCCATGAGAGATATCAT
Phenotype
MIM: 102770
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
AMPD1 PubMed Links

Gene Details

Gene
AMPD1
Gene Name
adenosine monophosphate deaminase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000036.2 1941 Missense Mutation TTA,TTT L631F NP_000027.2
NM_001172626.1 1941 Missense Mutation TTA,TTT L627F NP_001166097.1
Gene
DENND2C
Gene Name
DENN domain containing 2C
There are no transcripts associated with this gene.

View Full Product Details