Product Details

SNP ID
rs200320979
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:227815749 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTGCTTCCGTCATGGGCCCACT[C/T]GGGCCCTCTGCCCTGGGCCTTCTGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRSS38 PubMed Links

Gene Details

Gene
PRSS38
Gene Name
protease, serine 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_183062.2 33 Silent Mutation CTC,CTT L11L NP_898885.1
XM_011544175.2 33 Silent Mutation CTC,CTT L11L XP_011542477.1
XM_011544176.2 33 Silent Mutation CTC,CTT L11L XP_011542478.1

View Full Product Details