Product Details

SNP ID
rs200374238
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:153665264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCGAAGATTGGGTGGCACTGTT[A/G]TAATGAGAATCTTCACTGTAGCATC
Phenotype
MIM: 603181 MIM: 607007
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ILF2 PubMed Links

Gene Details

Gene
ILF2
Gene Name
interleukin enhancer binding factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267809.1 639 Missense Mutation ACA,ATA T140I NP_001254738.1
NM_004515.3 639 Missense Mutation ACA,ATA T178I NP_004506.2
Gene
SNAPIN
Gene Name
SNAP associated protein
There are no transcripts associated with this gene.

View Full Product Details