Product Details

SNP ID
rs200760827
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:204155017 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCTGCCTGGGTGGCAGAGGGCCT[C/T]AGCGGGCCAAGGCGAAGCCAATGCG
Phenotype
MIM: 609859 MIM: 179820
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ETNK2 PubMed Links

Gene Details

Gene
ETNK2
Gene Name
ethanolamine kinase 2
There are no transcripts associated with this gene.

Gene
REN
Gene Name
renin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000537.3 1264 Silent Mutation NP_000528.1

View Full Product Details