Product Details
- SNP ID
-
rs200482452
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:12775612 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTTCTGGGGCATATGGTCTGGAGC[A/T]TCTGCACTCTCCCCAGAGGCCCTGA
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PRAMEF12
PubMed Links
Gene Details
- Gene
- PRAMEF12
- Gene Name
- PRAME family member 12
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001080830.4 |
377 |
Silent Mutation |
GCA,GCT |
A119A |
NP_001074299.2 |
View Full Product Details