Product Details

SNP ID
rs200867914
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:247895644 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCATCCTACTGGGATTCTCTGAC[C/T]GACCCCATCTGGAGAGGATCCTCTT
Phenotype
MIM: 616729
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR2W3 PubMed Links

Gene Details

Gene
OR2W3
Gene Name
olfactory receptor family 2 subfamily W member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001957.2 58 Nonsense Mutation CGA,TGA R20* NP_001001957.2

View Full Product Details