Product Details

SNP ID
rs202113363
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:961673 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGACCCACGTGACGCTGCACGAC[A/G]TCGACCCTCAGGCCTTGGACCAGCT
Phenotype
MIM: 610770
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLHL17 PubMed Links

Gene Details

Gene
KLHL17
Gene Name
kelch like family member 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198317.2 1003 Missense Mutation ATC,GTC I138V NP_938073.1
XM_006710600.3 1003 Missense Mutation ATC,GTC I138V XP_006710663.1
XM_006710601.3 1003 Missense Mutation ATC,GTC I138V XP_006710664.1
Gene
NOC2L
Gene Name
NOC2 like nucleolar associated transcriptional repressor
There are no transcripts associated with this gene.

Gene
PLEKHN1
Gene Name
pleckstrin homology domain containing N1
There are no transcripts associated with this gene.

View Full Product Details