Product Details
- SNP ID
-
rs201867098
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:158563229 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTAGGGTAAAGGAGGGGTCCACAGA[C/T]GGCCAGGTAGCGATCATAGGCCATA
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR6P1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10489837] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR6P1
- Gene Name
- olfactory receptor family 6 subfamily P member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001160325.1 |
376 |
Missense Mutation |
ATC,GTC |
I126V |
NP_001153797.1 |
View Full Product Details