Product Details
- SNP ID
-
rs202157618
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
18
- Location
-
Chr.1:24057541 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCCCCTCACTTCCATGCGGTATCG[G/T]TCAAGGAAGGTGACAGGCTGGTCAT
- Phenotype
-
MIM: 616832
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MYOM3
PubMed Links
Gene Details
- Gene
- MYOM3
- Gene Name
- myomesin 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152372.3 |
4300 |
Silent Mutation |
GAG,GAT |
E1379D |
NP_689585.3 |
View Full Product Details