Product Details
- SNP ID
-
rs202203203
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:27967185 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAAGGCTAAGGATGAGGCTGCTTC[A/G]CCAGTGAAGGGATAGGTGAACGGCG
- Phenotype
-
MIM: 601655
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EYA3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs147164621] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EYA3
- Gene Name
- EYA transcriptional coactivator and phosphatase 3
There are no transcripts associated with this gene.
- Gene
- SMPDL3B
- Gene Name
- sphingomyelin phosphodiesterase acid like 3B
There are no transcripts associated with this gene.
- Gene
- XKR8
- Gene Name
- XK related 8
View Full Product Details