Product Details

SNP ID
rs200053406
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:25675388 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCCTTCCAGTTGAAGCCTCGCC[C/G]ACACTCCTTGCATACAAAAGGCTGC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF337 PubMed Links

Gene Details

Gene
ZNF337
Gene Name
zinc finger protein 337
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290261.1 1771 Missense Mutation CGG,GGG R634G NP_001277190.1
NM_015655.3 1771 Missense Mutation CGG,GGG R634G NP_056470.1
XM_005260702.3 1771 Missense Mutation CGG,GGG R602G XP_005260759.1
XM_006723558.3 1771 Missense Mutation CGG,GGG R634G XP_006723621.1
XM_011529219.2 1771 Missense Mutation CGG,GGG R634G XP_011527521.1
XM_017027802.1 1771 Missense Mutation CGG,GGG R602G XP_016883291.1
XM_017027803.1 1771 Missense Mutation CGG,GGG R602G XP_016883292.1
XM_017027804.1 1771 Missense Mutation CGG,GGG R569G XP_016883293.1
Gene
ZNF337-AS1
Gene Name
ZNF337 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details