Product Details

SNP ID
rs200121747
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:40688844 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCTGGCCCCATGCTCAGCTCCG[C/T]GGCCATCGCTGAAGCGAGGCGCAGC
Phenotype
MIM: 608968
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAFB PubMed Links

Gene Details

Gene
MAFB
Gene Name
MAF bZIP transcription factor B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005461.4 397 Missense Mutation ACG,GCG T3A NP_005452.2

View Full Product Details