Product Details

SNP ID
rs201783302
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:40688562 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGGCGTCAGGTTGAGCGCCTCGG[A/G]GTTCATCTGCTGGTAGTTGCTCGCC
Phenotype
MIM: 608968
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAFB PubMed Links

Gene Details

Gene
MAFB
Gene Name
MAF bZIP transcription factor B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005461.4 679 Missense Mutation CCC,TCC P97S NP_005452.2

View Full Product Details