Product Details

SNP ID
rs200631845
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:58693747 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTTCTGATCTAGCTCTGGCAGG[C/G]TGCCCTGAGCACGCTCAACCCCAAC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NPEPL1 PubMed Links

Gene Details

Gene
NPEPL1
Gene Name
aminopeptidase-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204872.1 199 Missense Mutation GCT,GGT A26G NP_001191801.1
NM_001204873.1 199 Missense Mutation GCT,GGT A6G NP_001191802.1
NM_024663.3 199 Missense Mutation GCT,GGT A54G NP_078939.3
Gene
STX16-NPEPL1
Gene Name
STX16-NPEPL1 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details