Product Details
- SNP ID
-
rs202089413
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:45479999 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACTTTCCCCAGCTCGGCCTCTGTC[A/G]GGACCAGTGCCAGGTGGACAGCCAG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
WFDC2
PubMed Links
Gene Details
- Gene
- WFDC2
- Gene Name
- WAP four-disulfide core domain 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006103.3 |
309 |
Missense Mutation |
CAG,CGG |
Q94R |
NP_006094.3 |
View Full Product Details