Product Details

SNP ID
rs200946979
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35174829 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAAGGCCCAGACACCAACACCAC[A/G]ATCATTCAGCTGCAGCCCTTGCAGG
Phenotype
MIM: 600646
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PROCR PubMed Links

Gene Details

Gene
PROCR
Gene Name
protein C receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006404.4 416 Silent Mutation ACA,ACG T66T NP_006395.2
XM_011528496.1 416 Silent Mutation ACA,ACG T66T XP_011526798.1

View Full Product Details