Product Details

SNP ID
rs201799359
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:25675197 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTTCATGCACAGTGAGGTCTGA[C/G]TTACTGGTATAGCCTCGCTTACACT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF337 PubMed Links

Gene Details

Gene
ZNF337
Gene Name
zinc finger protein 337
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290261.1 1962 Missense Mutation AAC,AAG N697K NP_001277190.1
NM_015655.3 1962 Missense Mutation AAC,AAG N697K NP_056470.1
XM_005260702.3 1962 Missense Mutation AAC,AAG N665K XP_005260759.1
XM_006723558.3 1962 Missense Mutation AAC,AAG N697K XP_006723621.1
XM_011529219.2 1962 Missense Mutation AAC,AAG N697K XP_011527521.1
XM_017027802.1 1962 Missense Mutation AAC,AAG N665K XP_016883291.1
XM_017027803.1 1962 Missense Mutation AAC,AAG N665K XP_016883292.1
XM_017027804.1 1962 Missense Mutation AAC,AAG N632K XP_016883293.1
Gene
ZNF337-AS1
Gene Name
ZNF337 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details