Product Details
- SNP ID
-
rs201842409
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:62126392 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGATGGCGCCCTACGGCCCGCTGG[C/T]GGCCAGCTCCCTGCTCAGCCAGCAG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LSM14B
PubMed Links
Gene Details
- Gene
- LSM14B
- Gene Name
- LSM family member 14B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_144703.2 |
583 |
Missense Mutation |
GCG,GTG |
A127V |
NP_653304.2 |
XM_005260302.3 |
583 |
Missense Mutation |
GCG,GTG |
A127V |
XP_005260359.1 |
XM_011528605.2 |
583 |
Missense Mutation |
GCG,GTG |
A153V |
XP_011526907.1 |
XM_011528606.2 |
583 |
Missense Mutation |
GCG,GTG |
A127V |
XP_011526908.1 |
XM_011528607.2 |
583 |
Missense Mutation |
GCG,GTG |
A153V |
XP_011526909.1 |
XM_011528608.2 |
583 |
Missense Mutation |
GCG,GTG |
A153V |
XP_011526910.1 |
XM_011528609.2 |
583 |
Missense Mutation |
GCG,GTG |
A153V |
XP_011526911.1 |
XM_011528611.2 |
583 |
Missense Mutation |
GCG,GTG |
A127V |
XP_011526913.1 |
XM_011528612.2 |
583 |
Missense Mutation |
GCG,GTG |
A8V |
XP_011526914.1 |
XM_011528613.2 |
583 |
Missense Mutation |
GCG,GTG |
A8V |
XP_011526915.1 |
XM_011528614.2 |
583 |
Missense Mutation |
GCG,GTG |
A8V |
XP_011526916.1 |
XM_011528615.2 |
583 |
Intron |
|
|
XP_011526917.1 |
XM_017027688.1 |
583 |
Missense Mutation |
GCG,GTG |
A153V |
XP_016883177.1 |
XM_017027689.1 |
583 |
Missense Mutation |
GCG,GTG |
A127V |
XP_016883178.1 |
XM_017027690.1 |
583 |
Intron |
|
|
XP_016883179.1 |
View Full Product Details