Product Details

SNP ID
rs200450712
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35547414 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGCTTCTCCCCTCCCCTTAGAG[C/G]TTGGGAAAGTCGAGGTGACGGTGTT
Phenotype
MIM: 616971
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ERGIC3 PubMed Links

Gene Details

Gene
ERGIC3
Gene Name
ERGIC and golgi 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015966.2 439 Missense Mutation CTT,GTT L124V NP_057050.1
NM_198398.1 439 Missense Mutation CTT,GTT L124V NP_938408.1

View Full Product Details