Product Details

SNP ID
rs199638108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35174910 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGCAGTCCTACCTGCTCCAGTT[C/G]CACGGCCTCGTGCGCCTGGTGCACC
Phenotype
MIM: 600646
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PROCR PubMed Links

Gene Details

Gene
PROCR
Gene Name
protein C receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006404.4 497 Missense Mutation TTC,TTG F93L NP_006395.2
XM_011528496.1 497 Missense Mutation TTC,TTG F93L XP_011526798.1

View Full Product Details