Product Details

SNP ID
rs199640143
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:40687990 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGTAGGCGTCTCTCTCGCGGGC[C/T]AGCCGGGACACCTCCTGCTTAAGCT
Phenotype
MIM: 608968
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAFB PubMed Links

Gene Details

Gene
MAFB
Gene Name
MAF bZIP transcription factor B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005461.4 1251 Silent Mutation CTA,CTG L287L NP_005452.2

View Full Product Details