Product Details

SNP ID
rs201143631
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44771461 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCGCGCCACATTTTGGACGCATC[C/T]ACGTTAGCTCCACTTTCGTCATTGG
Phenotype
MIM: 603124
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UBE2G2 PubMed Links

Gene Details

Gene
UBE2G2
Gene Name
ubiquitin conjugating enzyme E2 G2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202489.1 420 Silent Mutation GTA,GTG V68V NP_001189418.1
NM_003343.5 420 Silent Mutation GTA,GTG V138V NP_003334.2
NM_182688.2 420 Silent Mutation GTA,GTG V110V NP_872630.1
XM_017028469.1 420 Silent Mutation GTA,GTG V110V XP_016883958.1

View Full Product Details