Product Details

SNP ID
rs200201581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30282599 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCTGCAGCTGGGGCTGGGGCTGA[C/T]GTTGGTAGTTTCACAGACACTGAAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRTAP24-1 PubMed Links

Gene Details

Gene
KRTAP24-1
Gene Name
keratin associated protein 24-1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085455.2 360 Missense Mutation ATC,GTC I112V NP_001078924.1
Gene
KRTAP25-1
Gene Name
keratin associated protein 25-1
There are no transcripts associated with this gene.

View Full Product Details