Product Details

SNP ID
rs199992821
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30348520 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTAGACCAGGTTGCTGGGGTACGA[A/G]GAGCCACCACGGGAGTAGCCTGAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRTAP23-1 PubMed Links

Gene Details

Gene
KRTAP23-1
Gene Name
keratin associated protein 23-1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181624.1 87 Silent Mutation TCC,TCT S29S NP_853655.1

View Full Product Details