Product Details

SNP ID
rs200869965
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:32794167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGTGGCCCAGCGCAGATGTGTC[C/T]GTGAACCAGATGGTCAGATGGCCAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C21orf62 PubMed Links

Gene Details

Gene
C21orf62
Gene Name
chromosome 21 open reading frame 62
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162495.2 552 Silent Mutation ACA,ACG T85T NP_001155967.2
NM_001162496.2 552 Silent Mutation ACA,ACG T85T NP_001155968.2
NM_019596.5 552 Silent Mutation ACA,ACG T85T NP_062542.5
XM_011529653.2 552 Intron XP_011527955.1
Gene
C21orf62-AS1
Gene Name
C21orf62 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details