Product Details

SNP ID
rs199532937
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30598856 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCCATGGTCGCCGTAGTAGTTTC[C/T]GTAGTAGCTGCCGCACATCGTGATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRTAP22-1 PubMed Links

Gene Details

Gene
KRTAP22-1
Gene Name
keratin associated protein 22-1
There are no transcripts associated with this gene.

Gene
KRTAP22-2
Gene Name
keratin associated protein 22-2
There are no transcripts associated with this gene.

Gene
KRTAP6-2
Gene Name
keratin associated protein 6-2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181604.1 19 Missense Mutation AGA,GGA R7G NP_853635.1
Gene
KRTAP6-3
Gene Name
keratin associated protein 6-3
There are no transcripts associated with this gene.

View Full Product Details