Product Details

SNP ID
rs200167593
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50549568 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTACCGCCTGCTCAGGTACAGC[A/C]CCGTGAAGGATGCTTGGGACGAGTG
Phenotype
MIM: 615775
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KLHDC7B PubMed Links

Gene Details

Gene
KLHDC7B
Gene Name
kelch domain containing 7B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138433.3 1536 Missense Mutation ACC,CCC T468P NP_612442.2
Gene
SYCE3
Gene Name
synaptonemal complex central element protein 3
There are no transcripts associated with this gene.

View Full Product Details