Product Details

SNP ID
rs200831371
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37128205 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCTGAACTCCCCCTGCCCAGGA[A/G]GCCTGGACCAGGGGAAACTGACTCC
Phenotype
MIM: 146710
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL2RB PubMed Links

Gene Details

Gene
IL2RB
Gene Name
interleukin 2 receptor subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000878.3 1678 Missense Mutation CCT,CTT P516L NP_000869.1

View Full Product Details