Product Details

SNP ID
rs201929223
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:20426187 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACGCGGGGAGGGGTATCGCCTCCT[C/T]GGCGGAGGCTGGCGTGGTCAAGGGC
Phenotype
MIM: 613619
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SCARF2 PubMed Links

Gene Details

Gene
SCARF2
Gene Name
scavenger receptor class F member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153334.6 1894 Missense Mutation AAG,GAG K602E NP_699165.3
NM_182895.4 1894 Missense Mutation AAG,GAG K597E NP_878315.2
XM_017029065.1 1894 UTR 3 XP_016884554.1

View Full Product Details