Product Details

SNP ID
rs200409945
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:36284424 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACCTGGTCCTTGTACTGCTCGGC[A/G]TTCCTCCGCTCGTCATCCACCTGCA
Phenotype
MIM: 160775
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR6819 PubMed Links

Gene Details

Gene
MIR6819
Gene Name
microRNA 6819
There are no transcripts associated with this gene.

Gene
MYH9
Gene Name
myosin, heavy chain 9, non-muscle
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002473.5 5606 Silent Mutation AAC,AAT N1857N NP_002464.1
XM_011530197.2 5606 Silent Mutation AAC,AAT N1857N XP_011528499.1
XM_017028803.1 5606 Silent Mutation AAC,AAT N1857N XP_016884292.1
XM_017028804.1 5606 Silent Mutation AAC,AAT N1857N XP_016884293.1
XM_017028805.1 5606 Silent Mutation AAC,AAT N1857N XP_016884294.1
XM_017028806.1 5606 Silent Mutation AAC,AAT N1857N XP_016884295.1

View Full Product Details