Product Details
- SNP ID
-
rs200638885
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:99244120 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTATCCCAGGCACCGACAGGGTG[A/G]TACGTTTGTTTATCAAGCTAGAAAA
- Phenotype
-
MIM: 616547
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LYG2
PubMed Links
Gene Details
- Gene
- LYG2
- Gene Name
- lysozyme g2
View Full Product Details