Product Details

SNP ID
rs201026395
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:172427800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGTGCGTCCGCCCATGGCCGCCGC[C/G]GGGCAGCTGTGCTTGCTCTACCTGT
Phenotype
MIM: 147556
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ITGA6 PubMed Links

Gene Details

Gene
ITGA6
Gene Name
integrin subunit alpha 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000210.3 246 Silent Mutation GCC,GCG A4A NP_000201.2
NM_001079818.2 246 Silent Mutation GCC,GCG A4A NP_001073286.1
NM_001316306.1 246 Intron NP_001303235.1
XM_006712510.1 246 Silent Mutation GCC,GCG A4A XP_006712573.1
XM_006712511.1 246 Silent Mutation GCC,GCG A4A XP_006712574.1
XM_017004005.1 246 Intron XP_016859494.1
XM_017004006.1 246 Intron XP_016859495.1
XM_017004007.1 246 Intron XP_016859496.1
XM_017004008.1 246 Intron XP_016859497.1

View Full Product Details