Product Details

SNP ID
rs201217794
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:176123351 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTCCAAACGGACTGAGTGCTCC[G/T]TGGCCCGGGAGTCCCAGGGGAGCAG
Phenotype
MIM: 142984 MIM: 142985 MIM: 142982
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HOXD10 PubMed Links

Gene Details

Gene
HOXD10
Gene Name
homeobox D10
There are no transcripts associated with this gene.

Gene
HOXD8
Gene Name
homeobox D8
There are no transcripts associated with this gene.

Gene
HOXD9
Gene Name
homeobox D9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014213.3 667 Missense Mutation GTG,TTG V195L NP_055028.3
Gene
LOC100129455
Gene Name
uncharacterized LOC100129455
There are no transcripts associated with this gene.

View Full Product Details