Product Details

SNP ID
rs201288119
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:216259321 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCGGGCCTCTGCTCTCACACTGT[C/T]GTGACTCTCATGACCAGCTCTCGGC
Phenotype
MIM: 608208
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MARCH4 PubMed Links

Gene Details

Gene
MARCH4
Gene Name
membrane associated ring-CH-type finger 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020814.2 2991 Silent Mutation NP_065865.1

View Full Product Details